Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osteocalcin Mouse anti-Human, PE, Clone: R14-707, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564146

Catalog No. BDB564146


Add to cart

Description

Description

The R14-707 monoclonal antibody specifically recognizes human Osteocalcin. Osteocalcin (OC/OCN/OSTCN) is encoded by the BGLAP gene and is also known as Bone gamma-carboxyglutamic acid-containing protein. Osteocalcin is an approximately ~11 kDa protein that is secreted by osteoblasts. Its carboxylated form binds hydroxyapatite and thus it is abundantly found in bone extracellular matrix. Its uncarboxylated form serves as a hormone that can positively regulate glucose homeostasis. Serum osteocalcin has also been used as a biomarker for bone formation and turnover.

Specifications

Specifications

Osteocalcin
R14-707
BGLAP; BGP; Bone Gla protein; OC; OSTCN; PMF1
Affinity Purified
Human Osteocalcin Recombinant Protein
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit