Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osteocalcin Rabbit anti-Bovine, Canine, Human, Non-human primate, Polyclonal, Invitrogen™

Catalog No. PIPA185752
Change view
Click to view available options
Quantity:
250 μL
Catalog No. Quantity
PIPA185752 250 μL
1 options

Catalog No. PIPA185752

Supplier: Thermo Scientific PA185752

Rabbit Polyclonal Antibody

This antibody does not react with chicken, mouse or rat. This antibody is suitable for staining cultured human osteoblasts after fixation with 2% formaldehyde and permeabilization.

Osteocalcin belongs to the osteocalcin/matrix Gla-protein family and constitutes 1-2% of the total bone protein. It binds strongly to apatite and calcium. Gamma-carboxyglutamate residues are formed by vitamin K dependent carboxylation. These residues are essential for the binding of calcium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Osteocalcin
Applications Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Gene BGLAP
Gene Accession No. P02820, P81455, P02818
Gene Symbols BGLAP
Host Species Rabbit
Immunogen Human osteocalcin.
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 281646, 403762, 632
Target Species Bovine, Canine, Human, Non-human Primate
Content And Storage 4°C or -20°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.