Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osteoprotegerin/TNFRSF11B Mouse anti-Human, Clone: 69127, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $839.00

Specifications

Antigen Osteoprotegerin/TNFRSF11B
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Osteoprotegerin/TNFRSF11B Glu22-Leu401 Accession # O00300
Clone 69127
Applications ELISA
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB8051SP
View Documents
R&D Systems
MAB8051SP
25 ug
Each for $119.60
Add to cart
 
MAB8051100
View Documents
R&D Systems
MAB8051100
100 ug
Each for $377.00
Add to cart
 
MAB8051500
View Documents
R&D Systems
MAB8051500
500 ug
Each for $839.00
Add to cart
 
Description

Description

Osteoprotegerin/TNFRSF11B Monoclonal antibody MAB8051 Specifically detects Osteoprotegerin/TNFRSF11B in Human samples. It is validated for ELISA Capture
Specifications

Specifications

Osteoprotegerin/TNFRSF11B
69127
Unconjugated
O00300
Monoclonal
Primary
Detects human Osteoprotegerin/TNFRSF11B in ELISAs. It has been reported (1), and R&D Systems testing has confirmed, that this antibody shows strong cross-reactivity with an unrelated protein, human Carbonic Anhydrase II. In Western blots, this antibody does not cross-react with recombinant human (rh) 4-1BB, rhCD27, rhCD30, recombinant mouse (rm) CD30, rmCD40, rhDR3, rhDR6, rhFas, rmFas, rhGITR, rmGITR, rhHVEM, rhLT beta R, rhNGF R, rmOPG, rhTNF RI, rmTNF RI, rhTNF RII, or rmTNF RII.
Protein A or G purified from ascites
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Osteoprotegerin/TNFRSF11B Glu22-Leu401 Accession # O00300
ELISA
Purified
Mouse
IgG2a
4982
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit