Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

osteoclast stimulating factor 1 (B02), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human OSTF1 protein.

Manufacturer:  Abnova Corporation H00026578B02

Catalog No. 89-007-406


Add to cart

Description

Description

Osteoclast-stimulating factor-1 is an intracellular protein produced by osteoclasts that indirectly induces osteoclast formation and bone resorption (Reddy et al., 1998 [PubMed 10092216]).[supplied by OMIM

Sequence: MSKPPPKPVKPGEGGQVKVFRALYTFEPRTPDELYFEEGDIIYITDMSDTNWWKGTSKGRTGLIPSNYVAEQAESIDNPLHEAAKRGNLSWLRECLDNRVGVNGLDKAGSTALYWACHGGHKDIVEMLFTQPNIELNQQNKLGDTALHAAAWKGYADIVQLFLAKGARTDLRNIEKKLAFDMATNAACASLLKKKQGTDAVRTLSNAEDYLDDEDSD
Specifications

Specifications

osteoclast stimulating factor 1
Unconjugated
No additive
BC007459.1
FLJ20559|OSF|SH3P2|bA235O14.1
Mouse
50 uL
Signal Transduction
Y
Polyclonal
Human
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human OSTF1 protein.
OSTF1
Serum
OSTF1
OSTF1 (AAH07459.1, 1 a.a. ~ 217 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
26578
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit