Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Otubain-1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP149935

Catalog No. NBP149935


Add to cart

Description

Description

Otubain-1 Polyclonal antibody specifically detects Otubain-1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

Otubain-1
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Affinity Purified
OTUB1
The immunogen for this product maps to a region between residue 40 and 80 of human OTU Domain, Ubiquitin Aldehyde Binding 1 using the numbering given in entry NP_060140.2 (GeneID 55611).
Immunogen affinity purified
RUO
Primary
55611
Immunoprecipitation, Western Blot
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
Deubiquitinating enzyme OTUB1, EC 3.1.2, EC 3.4.19.12, FLJ20113, FLJ40710, hOTU1, MGC111158, OTB1MGC4584, OTU domain, ubiquitin aldehyde binding 1, OTU domain-containing ubiquitin aldehyde-binding protein 1, OTU1OTU-domain Ubal-binding 1, otubain-1, ubiquitin thioesterase OTUB1, ubiquitin-specific protease otubain 1, Ubiquitin-specific-processing protease OTUB1
Rabbit
IgG
0.1 ml
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only