Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OTUD3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524508

Catalog No. PIPA524508


Add to cart

Specifications

Specifications

OTUD3
Polyclonal
PBS with 0.09% sodium azide
Q5T2D3
DUBA4, RP11-460G22.1, OTU domain-containing protein 3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23252
Western Blot
Unconjugated
OTUD3
Liquid
OTUD3
KLH conjugated synthetic peptide between 192-220 amino acids from the central region of human OTUD3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit