Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-OTUD4, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136976

Catalog No. NBP136976


Add to cart

Description

Description

OTUD4 Polyclonal antibody specifically detects OTUD4 in Human, Mouse, Rat, Canine samples. It is validated for Peptide ELISA.
Specifications

Specifications

OTUD4
0.5 mg/mL
Peptide ELISA Detection limit 1:8000
Affinity Purified
OTUD4
Peptide with sequence C-SRSRDEGYQYHRNVR corresponding to C-Terminus (near) according to NP_001096123.1.
Immunogen affinity purified
RUO
Primary
54726
Canine, Human, Mouse, Rat
ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
DKFZp434I0721, HIV-1 induced protein HIN-1, HIV-1-induced protein HIN-1, HSHIN1, KIAA1046HIN1DUBA6, OTU domain containing 4, OTU domain-containing protein 4
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize reported isoforms 1 (NP_955356.1) and 3 (NP_001096123.1).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only