Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OTUD6B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525708

Catalog No. PIPA525708

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Deubiquitinating enzymes (DUBs; see MIM 603478) areproteases that specifically cleave ubiquitin (MIM 191339) linkages,negating the action of ubiquitin ligases. DUBA5 belongs to a DUBsubfamily characterized by an ovarian tumor (OTU) domain.
Specifications

Specifications

OTUD6B
Polyclonal
PBS with 0.09% sodium azide
Q8N6M0, Q8K2H2
DUBA5, DUBA-5, OTU domain-containing protein 6B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51633, 72201
Flow Cytometry, Western Blot
Unconjugated
OTUD6B
Liquid
OTUD6B
KLH conjugated synthetic peptide between 234-262 amino acids from the C-terminal region of human OTUD6B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit