Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OTX2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523406

Catalog No. PIPA523406


Add to cart

Description

Description

PA5-23406 is predicted to react with mouse due to 100% homology.

OTX2 is a 289 amino acid containing protein encoding a member of the bicoid sub-family of homeodomain-containing transcription factors. It has two known alternative splice variants with distinct isoforms though presence of other isoforms cannot be excluded. The protein is an early marker of endodermal differentiation of embryonic stem cells and may play a role in the development of the brain and the sensory organs. The protein may bind to BCD target sequence (BTS): 5’-TCTAATCCC-3’. Defects in OTX2 are the cause of microphthalmia syndromic type 5 (MCOPS5).
Specifications

Specifications

OTX2
Polyclonal
Unconjugated
OTX2
Liquid
OTX2
Synthetic peptide (27 PSVGYPATPRKQRRERT 43) of OTX2 protein.
Protein A
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P32243
CPHD6, MCOPS5, homeobox protein OTX2, orthodenticle homolog 2
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5015
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit