Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ovalbumin Mouse anti-Chicken, Clone: 8C6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen HYB0990902

Catalog No. HYB0990902

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

HYB 099-09-02 is specific for native chicken ovalbumin (OA). Cross-reactivity with egg albumin from other species has not been tested. HYB 099-09-02 has been successfully used in ELISA procedures. HYB 099-09-02 shows high affinity for OA in its native form. It does not recognize denatured OA. HYB 099-09-02 is not suitable for Western blot. The HYB 099-09-02 immunogen is purified chicken ovalbumin. Epitope specificity of HYB 099-09-02 differs from that of HYB 099-01-02 and HYB 099-02-02, as determined by inhibition ELISA. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL

Ovalbumin (OA) from chicken egg white, in which it makes up about 60% of solids. OA is a globular monomer of molecular mass ~45 kDa.
Specifications

Specifications

Ovalbumin
Monoclonal
0.96 mg/mL
PBS with 0.5M NaCl and 15mM sodium azide; pH 7.4
P01012
SERPINB14, Plakalbumin, Allergen Gal d 2
Mouse
IgG1, kappa
200 μL
4° C, store in dark
Primary
396058
ELISA
8C6
Unconjugated
OVAL
Liquid
OVAL
Purified chicken ovalbumin
Protein A
RUO
Antibody
Monoclonal
Chicken
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit