Learn More
OVCA1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA15857
Catalog No. 01-675-395
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Diphthamide is a unique posttranslationally modified histidine found only in translation elongation factor-2 (EEF2; MIM 130610). This modification is conserved from archaebacteria to humans and serves as the target for ADP-ribosylation and inactivation of EEF2 by diphtheria toxin (DT) and Pseudomonas exotoxin A. DPH1 is 1 of several enzymes involved in synthesis of diphthamide in EEF2 (Liu et al., 2004 [PubMed 15485916]).
Specifications
Specifications
OVCA1
Polyclonal
whole serum with 0.1% sodium azide
Q9BZG8
DPH2L, DPH2L1, FLJ33211, DPH1
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Western Blot
Unconjugated
DPH1
Liquid
DPH1
Human peptide (amino acid positions 2031) coupled to KLH
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1801