Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

candidate tumor suppressor in ovarian cancer 2, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human OVCA2 protein.

Manufacturer:  Abnova Corporation H00124641B01

Catalog No. 89-008-512


Add to cart

Description

Description

Sequence: MAAQRPLRVLCLAGFRQSERGFREKTGALRKALRGRAELVCLSGPHPVPDPPGPEGARSDFGSCPPEEQPRGWWFSEQEADVFCALEEPAVCRGLEESLGMVAQALNRLGPFDGLLGFSQGAALAALVCALGQAGDPRFPLPRFILLVSGFCPRGIGFKESILQRPLSLPSLHVFGDTDKVIPSQESVQLASQFPGAITLTHSGGHFIPAAAPQRQAYLKFLDQFAE
Specifications

Specifications

candidate tumor suppressor in ovarian cancer 2
Unconjugated
No additive
BC040696.2
OVCA2
OVCA2 (AAH40696.1, 1 a.a. ~ 227 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human OVCA2 protein.
OVCA2
Serum
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
124641
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit