Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse OX40 Ligand/TNFSF4 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 3 publications
$172.84 - $1184.00
Specifications
Antigen | OX40 Ligand/TNFSF4 |
---|---|
Clone | 182601 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Neutralization 0.01-0.05 ug/mL |
Applications | Western Blot, Neutralization |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1236SP
|
R&D Systems
MAB1236SP |
25 μg |
Each for $172.84
|
|
|||||
MAB1236100
|
R&D Systems
MAB1236100 |
100 μg |
Each for $532.44
|
|
|||||
MAB1236500
|
R&D Systems
MAB1236500 |
500 μg |
Each for $1,184.00
|
|
|||||
Description
OX40 Ligand/TNFSF4 Monoclonal specifically detects OX40 Ligand/TNFSF4 in Mouse samples. It is validated for Western Blot, Neutralization.Specifications
OX40 Ligand/TNFSF4 | |
LYOPH | |
Western Blot, Neutralization | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, TNFSF4, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1 | |
TNFSF4 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
182601 | |
Western Blot 1 ug/mL, Neutralization 0.01-0.05 ug/mL | |
Monoclonal | |
Rat | |
Mouse | |
P43488 | |
7292 | |
Mouse myeloma cell line NS0-derived recombinant mouse OX40 Ligand/TNFSF4 Gln49-Leu198 Accession # P43488 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse OX40 Ligand/TNFSF4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) APRIL, rhGITR Ligand, rhLIGHT, rhLT alpha 1/beta 2, rhLT alpha 2/beta 1, recombinant cotton rat TNF-alpha , recombinant mouse (rm) TNF-alpha , rmTRAIL, rhTRANCE, rmTRANCE, rmTWEAK, or rhVEGI is observed. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title