Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OX40L (CD252) Rat anti-Mouse, Clone: RM134L, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma 041470
Description
Anti-OX40L (CD252) Antibody, clone RM134L detects level of OX40L (CD252) and has been published and validated for use in Flow Cytometry and immunohistochemistry.Specifications
OX40L (CD252) | |
Monoclonal | |
Unconjugated | |
NP_033478 | |
Rat | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG2b κ |
Flow Cytometry, Immunohistochemistry | |
RM134L | |
Purified rat monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Tnfsf4, Ox40l, Txgp1l | |
Mouse OX40L cDNA-transfected NRK cells | |
100 μg | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only