Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-OX40L (CD252), PE-Cy7, Clone: RM134L,
Rat Monoclonal Antibody
Specifications
Antigen | OX40L (CD252) |
---|---|
Clone | RM134L |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE-Cyanine7 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF1446MI
|
MilliporeSigma™
MABF1446 |
25 μg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Specifically detects OX40L (CD252) Clone: RM134L in Mouse samples, and it is validated for Flow Cytometry
The RM134L antibody recognizes CD252, also known as OX-40 Ligand or CD134 Ligand, a member of the TNF superfamily that is present on the surface of antigen presenting cells and activated B lymphocytes. The OX-40 Ligand interacts with OX-40 (CD134) which is expressed primarily on activated T cells. This costimulatory interaction leads to increased proliferation and IL-2 production responses of activated T cells, and at the same time enhances proliferation and immunoglobulin secretion by activated B cells.The RM134L antibody is useful for flow cytometric detection of CD252 on stimulated mouse splenocytes.Specifications
OX40L (CD252) | |
Flow Cytometry | |
PE-Cyanine7 | |
Rat | |
Inflammation, Immunology | |
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
Tnfsf4, Ox40l, Txgp1l | |
IgG2b κ | |
Purified by size-exclusion chromatography |
RM134L | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
P43488 | |
Proprietary immunogen | |
Primary | |
2°C to 8°C for one year from date of shipment protected from light; Do not freeze |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title