Learn More
Apolipoprotein B Mouse anti-Human, Clone: 15G2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen HYB2620302
Catalog No. HYB2620302
Add to cart
Description
Description
HYB 262-03-02 reacts with MDA-oxidized LDL but not with native LDL. HYB 262-03-02 has been successfully used in ELISA and Western blotting techniques. HYB 262-03-02 reacts with MDA-oxidized LDL in ELISA and in SDS-PAGE immunoblotting. The HYB 262-03-02 immunogen is low-density lipoprotein purified from human plasma and oxidized with malondialdehyde (MDA). NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Apolipoprotein B (apo B) in human plasma is a major protein of low density lipoproteins (LDL) with a molecular mass of approximately 260-500 kDa. Apolipoprotein B binds to specific receptors on cell membranes and is involved in removal of LDL and very low density lipoprotein (VLDL) cholesterol from circulation. Low-density lipoprotein (LDL) is the carrier protein for cholesterol in the blood. LDL binds to its receptor on the capillary walls and thereby mediates the uptake and clearance of cholesterol from the circulation. In atherosclerotic lesions oxidatively modified LDL is found and oxidized LDL is specifically recognized and ingested by macrophages via scavenger receptor A and CD36. Oxidized LDL may be a marker of atherosclerosis but the precise changes in oxidized LDL are not well described. MDA-oxidized LDL appear to be different from LDL oxidized by other means. Apolipoprotein B is mostly synthesized in the liver, and is a major apolipoprotein of very low density, intermediate density and low density lipoproteins (LDL) as well as being a major component of lipoprotein (a). Apolipoprotein B is a ligand for the LDL receptor and elevated levels are associated with premature atherosclerosis. Normal plasma apolipoprotein B levels are around 800mg/L. Apolipoprotein B occurs in plasma as two main isoforms, apoB-48 and apoB-100. The intestinal and the hepatic forms of Apolipoprotein B are encoded by a single gene from a single, very long mRNA. The two isoforms share a common N-terminal sequence. The shorter apoB-48 protein is produced after RNA editing of the apoB-100 transcript at residue 2180 (CAA->UAA), resulting in the creation of a stop codon, and early translation termination. Mutations in the Apolipoprotein B gene or its regulatory region cause hypobetalipoproteinemia, normotriglyceridemic hypobetalipoproteinemia, and hypercholesterolemia due to ligand-defective Apolipoprotein B, diseases affecting plasma cholesterol and Apolipoprotein B levels.
Specifications
Specifications
Apolipoprotein B
Monoclonal
1 mg/mL
PBS with 15mM sodium azide; pH 7.4
P04114
Ox-LDL receptor 1, Lectin-type oxidized LDL receptor 1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, LOX-1
Mouse
IgG2a, kappa
200 μL
4° C, store in dark
Primary
338
ELISA, Western Blot
15G2
Unconjugated
APOB
Liquid
APOB
Low-density lipoprotein purified from human plasma and oxidized with malondialdehyde (MDA).
Protein A
RUO
Antibody
Monoclonal
Human