Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OXPAT Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB11060511SS
Description
OXPAT Polyclonal specifically detects OXPAT in Human, Mouse samples. It is validated for Western Blot, Simple Western.Specifications
OXPAT | |
Polyclonal | |
Unconjugated | |
PBS and 30% Glycerol with 0.1% Sodium Azide | |
PLIN5 | |
A synthetic peptide made to a C-terminal region of the mouse OXPAT protein, within residues 400-463. [UniProt# Q8BVZ1] | |
Affinity Purified | |
RUO | |
Primary | |
Human and mouse. Immunogen sequence has 89% homology to rat and 75% homology to bovine. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot | |
1.01 mg/mL | |
Western Blot 0.5-2 ug/ml, Simple Western 10 ug/ml | |
lipid storage droplet protein 5, LSDA5, LSDP5, MLDP, OXPAT, perilipin 5, perilipin-5 | |
Rabbit | |
50 kDa | |
0.025 mL | |
Diabetes Research, Lipid and Metabolism, Lipid Droplets | |
440503 | |
Human, Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction