Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OXPAT Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA146216

Catalog No. PA146216


Add to cart

Description

Description

The target sequence has 89% sequence homology with rat and 75% sequence homology with bovine. Suggested positive control: human and mouse heart lysate.

OXPAT is a novel PAT-1 protein that is expressed in highly oxidative tissues. Lipid droplet proteins of the PAT family regulate cellular neutral lipid stores. OXPAT is induced by physiological (fasting), pathophysiological (insulin deficiency), pharmacological (peroxisome proliferator-activated receptor [PPAR] agonists), and genetic (muscle-specific PPAR alpha overexpression) disturbances that increase fatty acid utilization. It is suggested that OXPAT is a marker for PPAR activation and fatty acid oxidation. As well, it may contribute to adaptive responses to the fatty acid responsibility that accompanies fasting, insulin deficiency, and overnutrition, responses that are defective in obesity and type 2 diabetes.
Specifications

Specifications

OXPAT
Polyclonal
Unconjugated
PLIN5
Liquid
PLIN5
C-terminal region of the mouse OXPAT protein, with in residues 400-463 [UniprotNo. Q8BVZ1].
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
1.01 mg/mL
PBS with 30% glycerol and 0.1% sodium azide
Q00G26, Q8BVZ1
LSDA5, LSDP5, MLDP, OXPAT
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
440503, 66968
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit