Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OXR1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526599

Catalog No. PIPA526599

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

OXR1 may be involved in protection from oxidative damage.
Specifications

Specifications

OXR1
Polyclonal
PBS with 0.09% sodium azide
Q8N573
oxidation resistance protein 1, putative protein product of Nbla00307
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55074
Western Blot
Unconjugated
OXR1
Liquid
OXR1
KLH conjugated synthetic peptide between 268-296 amino acids from the N-terminal region of human OXR1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit