Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OXR1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen PA526600
Description
This antibody is predicted to react with rat based on sequence homology.
OXR1 may be involved in protection from oxidative damage.Specifications
OXR1 | |
Polyclonal | |
Liquid | |
OXR1 | |
oxidation resistance protein 1, putative protein product of Nbla00307 | |
OXR1 | |
KLH conjugated synthetic peptide between 596-625 amino acids from the central region of human OXR1 | |
Polyclonal | |
Antigen affinity chromatography | |
RUO | |
Human |
Western Blot | |
Unconjugated | |
PBS with 0.09% sodium azide | |
Q8N573 | |
55074 | |
Rabbit | |
IgG | |
Primary | |
400 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Antibody |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title