Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-OXTR, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128487

Catalog No. NBP128487


Add to cart

Description

Description

OXTR Polyclonal antibody specifically detects OXTR in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications

Specifications

OXTR
0.5 mg/mL
Western Blot 1-3ug/ml, Immunohistochemistry 2 - 4 ug/ml, Immunocytochemistry/Immunofluorescence 5 - 10 ug/ml, Immunohistochemistry-Paraffin 2-4 ug/ml, Peptide ELISA detection limit 1:16000
NP_000907.2
OT-R, oxytocin receptor
Goat
IgG
0.1 mg
Cancer, GPCR, Neuroscience, Neurotransmission
Primary
5021
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
OXTR
Peptide with sequence C-RRLGETSASKKSN corresponding to C-Terminus according to NP_000907.2.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only