Learn More
anti-P-Cadherin, PE, Clone: 106020, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB761P
Catalog No. FAB761P
Add to cart
This item is not returnable. View return policy
Description
Description
Applications:
Flow Cytometry
Specifications
Specifications
P-Cadherin
106020
Flow Cytometry 10 uL/10^6 cells
P10287
CAD3, cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDH3, CDHP, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCAD, PCadherin, PCADP-cadherin, Placental cadherin
Rat
IgG2a
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects mouse P-Cadherin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) N-Cadherin, recombinant mouse (rm) VE-Cadherin, rhCadherin-8, or rhCadherin-17 is observed.
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CDH3
Mouse myeloma cell line NS0-derived recombinant mouse P-Cadherin Glu100-Gly647 Accession # P10287
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1001
Human, Mouse