Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-P LEUCOPUSIGG (H+L) AB AP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52200382
Affinity purified polyclonal antibody to white-footed mouse IgG, both heavy and light chains (whole IgG) made in Goat and labeled with alkaline phosphatase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-54