Learn More
anti-P-Selectin/CD62P, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP150764
Catalog No. NBP150764
Add to cart
Description
Description
P-Selectin/CD62P Polyclonal antibody specifically detects P-Selectin/CD62P in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
P-Selectin/CD62P
0.5 mg/mL
Western Blot 0.5 - 4 ug/mL, Immunohistochemistry
Q01102
CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62)
Rabbit
IgG
Affinity purified
RUO
Primary
6403
Immunohistochemistry, Western Blot
Unconjugated
PBS (pH 7.2), 0.5% BSA, 30% Glycerol with 0.01% Thimerosal
Affinity Purified
SELP
Synthetic peptide surrounding amino acid 756 of mouse CD62P
91 kDa
0.1 mg
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat