Learn More
p107 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific PA129140
Catalog No. PIPA129140
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
PA1-29149 detects ARC from human samples. PA1-29149 has been successfully used in immunohistochemistry (paraffin tissue) applications. The PA1-29149 immunogen is: Synthetic peptide corresponding to the C-terminus of Human A
p107 is related both in structure and function to the retinoblastoma tumor suppressor protein and p130 (collectively known as pocket proteins) and are known to regulate the activity of E2F transcription factors. E2F transcription factors regulate the expr
TRUSTED_SUSTAINABILITY
Specifications
Specifications
p107
Polyclonal
Unconjugated
Q08828, O88444
ADCY1
Synthetic peptide corresponding to C-terminus of human p107 protein
500 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Western Blot
1 mg/mL
ADCY1
Retinoblastoma related protein
Rabbit
Protein A
RUO
107, 305509, 432530
4° C, do not freeze
Liquid
Recommendations
Recommendations