Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p115 Mouse, Unlabeled, Clone: 15, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612261

 View more versions of this product

Catalog No. BDB612261


Add to cart

Description

Description

Maturation and post translational modification of proteins occurs after their biosynthesis at the endoplasmic reticulum and their transport through the Golgi apparatus. The process involves the transport of vesicles carrying the proteins through a vectorial process of vesicle budding and fusion from the cis-compartment to the medial-compartment and the trans-compartment of the Golgi apparatus. p115 is a 959 amino acid protein located at the Golgi apparatus that, with the NEM-sensitive fusion protein and the soluble NSF attachment protein (SNAP), is required for vesicle transport from the cis-compartment to the medial-compartment. p115 protein is related to the yeast Uso1p essential for the vesicular transport from the endoplasmic reticulum to the Golgi. Native p115 appears to be a homo-oligomer, with two globular heads and a tail that resemble the overall structure of myosin. p115 is extracted from the Golgi apparatus with high salt or high pH, indicative of a membrane associated protein. p115 interacts with the golgi matrix protein GM130 but this interaction is disrupted by the Golgi fragmentation during mitosis and the phosphorylation of GM130.

Immunofluorescence, Western Blotting

Specifications

Specifications

p115
15
Unconjugated
Affinity Purified
Rat p115 aa. 843-955
150μg
Cell Biology
Primary
Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.