Learn More
VDP Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA1006
Catalog No. PA1006
Add to cart
Description
Description
PA1-006 detects p115 in rat and mouse samples. PA1-006 has been successfully used in Western blot procedures. By Western blot, this antibody detects a ~115 kDa protein representing p115 in rat and mouse liver samples. The PA1-006 immunogen is a synthetic peptide corresponding to residues M(1) N F L R G V M G G Q S A G P Q H(17) of rat p115. This sequence is conserved across many species including mouse and human. PA1-006 immunizing peptide (Cat. No. PEP-276) is available for use in neutralization and control experiments.
p115 (Vesicle docking protein p115) is a peripheral membrane protein that is located on the Golgi complex. p115 exists as a homodimer with two globular heads, an extended coiled-coil tail, followed by an acidic domain at the extreme C terminus. p115 is homologous to a yeast protein, Uso1p, which is required for ER to Golgi transport. p115 likely plays an important role in vesicle transportation from the ER to the cis-Golgi comparments.
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
VDP
Polyclonal
Unconjugated
USO1
Liquid
USO1
Synthetic peptide corresponding to residues M(1) N F L R G V M G G Q S A G P Q H(17) of rat p115.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
O60763, Q9Z1Z0, P41542
TAP, Transcytosis associated protein, Vdp, General vesicular transport factor p115, Vesicle docking protein
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56041, 56042, 8615