Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p120 Catenin Mouse anti-Human, Mouse, Rat, Clone: 15D2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051567
Description
Use Anti-p120 Catenin Antibody, clone 15D2 (mouse monoclonal antibody) validated in western blot, immunocytochemistry, FC to detect p120 Catenin also known as Cadherin-associated Src substrate ,catenin (cadherin-associated protein) delta 1.Specifications
p120 Catenin | |
Monoclonal | |
Unconjugated | |
O60716 | |
Mouse | |
Protein G chromatography | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Flow Cytometry, Immunocytochemistry, Western Blot | |
15D2 | |
Purified mouse monoclonal in 0.1M Tris-Glycine (pH 7.4), 150mM NaCl and 0.05% sodium azide. | |
CAS , CTNND, KIAA0384, P120CAS, p120, P120CTN, p120(cas), p120(ctn), p120cas , p120ctn | |
Recombinant fusion protein derived from the C-terminus of murine p120 | |
100 μg | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only