Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p120 Catenin (pY280) Mouse, Unlabeled, Clone: 18, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612538

Catalog No. BDB612538


Add to cart

Description

Description

The membrane associated protein pp120 Src substrate (p120 Catenin, p120cas) was identified as a tyrosine kinase substrate that is phosphorylated in Src transformed cells or in response to growth factor stimulation. It shares structural similarity with the Drosophila Armadillo protein and the vertebrate β-catenin and γ-catenin proteins. This similarity is evidenced by its characteristic Arm domain that is composed of 42-amino acid motif repeats. In the cell, p120 Catenin is localized to the E-Cadherin/catenins cell adhesion complex. Like β- and γ-catenin, p120 Catenin directly associates with the cytoplasmic C-terminus of E-Cadherin via its Arm domain and may similarly interact with other Cadherins. It exists as four isoforms that range in size from 90-115kDa. Expression of these isoforms is heterogeneous in human carcinomas, suggesting that altered pp120 expression contributes to malignancy due to loss of functional cell adhesions. Multiple tyrosine residues (Y96, Y112, Y228, Y280, Y257, Y291, Y296, and Y302) in p120 Catenin are phosphorylated by Src and these phosphorylations
may facilitate interaction with PTP1C/SHP-1 in response to EGF stimulation. Thus, p120 Catenin is an Arm domain protein that interacts with both cell adhesion molecules, such as cadherins and cell signaling molecules, such as PTP1C.

Immunofluorescence, Western Blotting

Specifications

Specifications

p120 Catenin (pY280)
18
Unconjugated
Affinity Purified
Human p120 Catenin (pY280) aa. 1-911
50μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit