Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p14ARF Rabbit anti-Human, Polyclonal, Invitrogen™

Catalog No. PIPA128218
Change view
Click to view available options
Quantity:
7 mL
Catalog No. Quantity
PIPA128218 7 mL
1 options

Catalog No. PIPA128218

Supplier: Thermo Scientific PA128218

Rabbit Polyclonal Antibody

PA1-28252 detects CaMKI from human, mouse, rat samples. PA1-28252 has been successfully used in immunoprecipitation, Western blot applications. The PA1-28252 immunogen is: Fusion protein from the N-terminal of Ca-Calmodulin kinase.

This gene generates several transcript variants which differ in their first exons. At least three alternatively spliced variants encoding distinct proteins have been reported, two of which encode structurally related isoforms known to function as inhibitors of CDK4 kinase. The remaining transcript includes an alternate first exon located 20 Kb upstream of the remainder of the gene; this transcript contains an alternate open reading frame (ARF) that specifies a protein which is structurally unrelated to the products of the other variants. This ARF product functions as a stabilizer of the tumor suppressor protein p53 as it can interact with, and sequester, MDM1, a protein responsible for the degradation of p53. In spite of the structural and functional differences, the CDK inhibitor isoforms and the ARF product encoded by this gene, through the regulatory roles of CDK4 and p53 in cell cycle G1 progression, share a common functionality in cell cycle G1 control. This gene is frequently mutated or deleted in a wide variety of tumors, and is known to be an important tumor suppressor gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p14ARF
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Gene CDKN2A
Gene Accession No. P42771
Gene Alias ARF, CDK4I, CDKN2, CMM2, INK4, INK4a, MLM, MTS1, TP16, p14, p14ARF, p16, p16INK4, p16INK4a, p19, p19Arf
Gene Symbols CDKN2A
Host Species Rabbit
Immunogen Synthetic peptidecorresponding to the Human C terminal region.
Purification Method Antigen affinity chromatography
Quantity 7 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1029
Target Species Human
Content And Storage 4° C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.