Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

regulation of nuclear pre-mRNA domain containing 1A, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human RPRD1A protein.

Manufacturer:  Abnova Corporation H00055197B01

Catalog No. 89-007-816


Add to cart

Description

Description

P15RS is upregulated in cells overexpressing cyclin-dependent kinase inhibitor p15(INK4b) (CDKN2B; MIM 600431) and may have a role in cell cycle regulation (Liu et al., 2002 [PubMed 12470661]).[supplied by OMIM

Sequence: MSAFSEAALEKKLSELSNSQQSVQTLSLWLIHHRKHSRPIVTVWERELRKAKPNRKLTFLYLANDVIQNSKRKGPEFTKDFAPVIVEAFKHVSSETDESCKKHLGRVLSIWEERSVYENDVLEQLKQALYGDKKPRKRTYEQIKVDENENCSSLGSPSEPPQTLDLVRALQDLENAASGDAAVHQRIASLPVEVQEVSLLDKITDKESGERLSKMVEDACMLLADYNGRLAAEIDDRKQLTRMLADFLRCQKEALAEKEHKLEILDMGVSSNDVVTISIFFLLKINIKP
Specifications

Specifications

regulation of nuclear pre-mRNA domain containing 1A
Unconjugated
No additive
BC010136.2
FLJ10656|HsT3101|MGC19513|P15RS
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
55197
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human RPRD1A protein.
RPRD1A
Serum
RPRD1A
RPRD1A (AAH10136.1, 1 a.a. ~ 289 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit