Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p16INK4 with control Mouse anti-Human, Unlabeled, Clone: G175-405, BD
Mouse Monoclonal Antibody
$254.00
Specifications
Antigen | p16INK4 with control |
---|---|
Clone | G175-405 |
Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB551153
|
BD Biosciences
551153 |
50 μg |
Each of 1 for $254.00
|
|
|||||
Description
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting
Specifications
p16INK4 with control | |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
RUO | |
IgG1 | |
Affinity Purified |
G175-405 | |
Monoclonal | |
Mouse | |
Human | |
Primary |
For Research Use Only.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title