Learn More
p19 INK4d Mouse anti-Human, Clone: DCS100, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA112304
Catalog No. 01-670-997
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
p19 / INK 4d is a specific inhibitor of cdk 4 / cdk 6 and is periodically expressed during cell cycle. It is maximally induced as cells enter S-phase.
Specifications
Specifications
p19 INK4d
Monoclonal
Unconjugated
CDKN2D
Liquid
CDKN2D
His-tagged recombit full length p19 protien.
Protein A/G
RUO
Antibody
Monoclonal
Human
Immunohistochemistry, Immunoprecipitation, Western Blot
DCS100
PBS with 0.08% sodium azide
P55273
Cyclin-dependent kinase 4 inhibitor A, CDK4I, p16-INK4, p16-INK4a, p16INK4A Multiple tumor suppressor 1, MTS1
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
1032