Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p190 Mouse, Unlabeled, Clone: 30, BD

Mouse Monoclonal Antibody

$276.00 - $508.00

Specifications

Antigen p190
Clone 30
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610149
View Documents
BD Biosciences
610149
50 μg
Each for $276.00
Add to Cart
 
BDB610150
View Documents
BD Biosciences
610150
150 μg
Each for $508.00
Add to Cart
 
Description

Description

GTPase activating proteins (GAPs) stimulate the GTP-hydrolyzing activity of GTPases, such as p21[ ras]. Ras-GAP (120kDa) is perhaps the best characterized GAP. p190 is a Ras-GAP associated protein that is tyrosine phosphorylated in transformed and growth factor-stimulated cells. Ras-GAP and p190 are targets of oncoproteins and growth factor receptors. Recombinant p190 enhances GTPase activity of the rho family of GTPases. The complete sequence of p190 exhibits three major regions: an N-terminal region with homology to all GTPases, a middle region homologous to transcriptional repressors, and a C-terminal region the is similar to n-chimerin and other small GTPases.

Specifications

Specifications

p190
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
30
Western Blot
Unconjugated
RUO
Canine, Chicken, Frog, Human, Mouse, Rat
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Store undiluted at -20°C.
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit