Learn More
p19ARF Rat anti-Mouse, Clone: 5-C3-1, MilliporeSigma™
Rat Monoclonal Antibody
Manufacturer:  MilliporeSigma 05929
Catalog No. 05-929-MI
Add to cart
Description
Description
Anti-p19ARF Antibody, clone 5-C3-1 is a high quality Rat Monoclonal Antibody for the detection of p19ARF and has been validated in immunoprecipitation, western blotting and immunohistochemistry.
Specifications
Specifications
p19ARF
5-C3-1
NM_000077.3; NM_058195.2; NM_058197.3
CDKN2A, CDKN2, P16, CMM2, ARF, p19ARF, p14ARF, TP16, p16, MTS1, INK4a, MLM, p16INK4a, INK4, p16-INK4a, p14, p16-INK4, p16INK4A, p19, p16INK4, CDK4I
KLH-conjugated, synthetic peptide corresponding to amino acids 54-75 (RRGPHRNPG-PGDDDGQRSRSSS) of mouse p19ARF. The epitope is mapped to amino acids 62-75. Clone 5-C3-1.
1mL
Primary
Murine
Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
Supernatant
Rat
IgG2b
−20°C for 24 months.
Monoclonal