Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP2 Mouse, Alexa Fluor 647, Clone: FOXP2-73A/8, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564463

Catalog No. BDB564463


Add to cart

Description

Description

The FOXP2-73A/8 monoclonal antibody specifically binds to Forkhead box protein P2 (FoxP2). FoxP2 is also known as TNRC10 (Trinucleotide repeat-containing gene 10 protein), CAGH44 (CAG repeat protein 44), and SPCH1 (Speech and Language Disorder 1). FoxP2 is a member of the forkhead/winged-helix (FOX) family of transcription factors. Normal FOXP2 gene expression is associated with the proper development of cardiovascular, gastrointestinal, and neural tissues including the speech and language regions of the brain. FoxP2 protein has been detected in most normal tissues including epithelial and endothelial cells but not cells of hematopoietic origin. Although not normally detected in lymphocytes and plasma cells, FoxP2 protein was reportedly expressed in a small fraction of plasma cells from reactive human tonsils. Aberrant FOXP2 gene expression has been associated with abnormal plasma cell generation and may play a role in multiple myeloma and other hematological malignancies. The FOXP2-73A/8 antibody detects human and mouse FoxP2 and does not crossreact with other members of the FOXP family (FoxP1, FoxP3 or FoxP4).

Specifications

Specifications

FoxP2
FOXP2-73A/8
Alexa Fluor 647
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
IgG1, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
TNRC10; SPCH1; CAGH44
Affinity Purified
Human FoxP2 Recombinant Protein
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Human, Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit