Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p21-Arc Mouse, FITC, Clone: 26, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612236

Catalog No. BDB612236


Add to cart

Description

Description

Cellular morphology, adhesion, and motility occur through the reorganization of the actin cytoskeleton. This reorganization of actin filaments occurs through the interactions between actin and actin binding proteins. Actin-binding proteins regulate the polymerization and depolymerization of actin, connect actin-based structures to membranes and to other cytoskeletal elements, power the movement of actin filaments, and cross-link actin filaments into bundles. Actin related proteins (Arp) 2/3 complex is an actin polymerization inducing complex that includes Arp2, Arp3, p41-Arc, p34-Arc, p21-Arc, p20 Arc, and p16-Arc. The Arp2 and Arp3 subunits may nucleate actin polymerization, while the p41-Arc subunit is a WD repeat-containing protein that may regulate both the activity and localization of the Arp2/3 complex. Arp3, p34-Arc, and p21-Arc are localized to the lamellipodia of stationary and locomoting fibroblasts. Both WASP and Abp1p are acidic sequence-containing proteins that activate the Arp2/3 complex. However, WASP binds actin monomers, while the endocytosis-related Abp1p protein binds actin filaments. Thus, Arp2/3 complex may regulate actin polymerization in specific cell locations through interaction with actin binding Arp2/3 activators.

Immunofluorescence

Specifications

Specifications

p21-Arc
26
FITC
Affinity Purified
Human p21-Arc aa. 10-118
50μg
Cell Biology
Primary
Canine, Chicken, Human, Murine, Rat
Immunofluorescence
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit