Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2X3 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124625

Catalog No. PA124625


Add to cart

Description

Description

PA1-24625 detects P2X3 from human, rat, and non-human primate samples. PA1-24625 is expected to cross react with mouse (100% conserved) due to sequence homology. PA1-24625 has been successfully used in immunohistochemistry (frozen), and Western blot procedures. The PA1-24625 immunogen is a synthetic peptide corresponding to residues V(383) E K Q S T D S G A Y S I G H(397) of C-terminus of rat P2X3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel and may transduce ATP-evoked nociceptor activation. Mouse studies suggest that this receptor is important for peripheral pain responses, and also participates in pathways controlling urinary bladder volume reflexes. It is possible that the development of selective antagonists for this receptor may have a therapeutic potential in pain relief and in the treatment of disorders of urine storage.
Specifications

Specifications

P2X3
Polyclonal
whole serum with 0.05% sodium azide
P56373, Q3UR32, P49654
MGC129956, P2X3, p2xr3
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Non-human primate, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
Unconjugated
P2RX3
Liquid
P2RX3
Synthetic peptide corresponding to residues V(383) E K Q S T D S G A Y S I G H(397) of C-terminus of rat P2X3.
50 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
228139, 5024, 81739
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit