Learn More
MilliporeSigma™ anti-P2X4 Receptor (373-388), Polyclonal
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma™ PC376100UG
Catalog No. PC376100UG
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifically detects P2X4 Receptor (373-388) Clone: in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry (Frozen), Immuno Blotting
Extracellular ATP acts as a signaling molecule to mediate a wide variety of physiological responses through interactions with the P2 family of receptors. P2 receptors have been classified into two groups, the P2X family of ligand-gated ion channels, and the P2Y family of metabotropic G-protein coupled receptors. To date, seven P2X receptor subtypes and eight different P2Y receptor subtypes have been identified. The P2X receptor subunits exhibit approximately 35%-48% identity with each other at the amino acid level, and have been shown to form both homo- and hetereromultimeric complexes. The various P2X receptor subunits exhibit a broad pattern of distribution, being localized to different types of immune cells, glands, smooth muscle, skeletal muscle, and the central and peripheral nervous systems. The P2X4 receptor subunit is predominantly expressed in the central nervous system and has a widespread distribution as determined byin situ hybridization and immunohistochemistry.
Specifications
Specifications
P2X4 Receptor (373-388)
Unconjugated
purified
a synthetic peptide (KYVEDYEQGLSGEMNQ) corresponding to amino acids 373-388 of rat P2X4 receptor
100μg
2°C to 8°C
Polyclonal
Human, Murine, Rat
Immuno Blotting, Immunoblot, Immunohistochemistry (Frozen)
In 0.05M sodium phosphate buffer, 0.2% gelatin.
Rabbit
IgG
RUO
Primary
Recognizes the ∽65kDa P2X4 receptor protein in mouse brain.