Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

anti-P2Y1/P2RY1, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP120959

Catalog No. NBP120959



Description

Description

P2Y1/P2RY1 Polyclonal antibody specifically detects P2Y1/P2RY1 in Human samples. It is validated for Peptide ELISA.
Specifications

Specifications

P2Y1/P2RY1
0.5 mg/mL
Peptide ELISA Detection limit 1:8000
NP_002554.1
ATP receptor, P2 purinoceptor subtype Y1, P2Y purinoceptor 1, P2Y1platelet ADP receptor, Purinergic receptor, purinergic receptor P2Y, G-protein coupled, 1
Goat
IgG
0.1 mg
Apoptosis, Cytokine Research, GPCR, Plasma Membrane Markers
Primary
5028
Human
ELISA
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
P2RY1
Peptide with sequence C-RSEANLQSKSED corresponding to C-Terminus according to NP_002554.1.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only