Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-P2Y2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB11039032

 View more versions of this product

Catalog No. NB11039032

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

P2Y2 Polyclonal antibody specifically detects P2Y2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

P2Y2
1 mg/mL
Western Blot 1:500-1:1000, Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 1:100 - 1:200
P41231
ATP receptor, HP2U, MGC20088, MGC40010, P2RU1, P2U nucleotide receptor, P2U purinoceptor 1, P2U1, P2UP2UR, P2Y purinoceptor 2, P2Y2P2Y2R, Purinergic receptor, purinergic receptor P2Y, G-protein coupled, 2, purinergic receptor P2Y2, purinoceptor P2Y2
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS with 0.05% Sodium Azide
Affinity Purified
P2RY2
A synthetic peptide made to a C-terminal portion of the human P2Y2 protein (between animo acids 300-377) [UniProt P41231].
42.3 kDa
0.1 mg
GPCR, Neuroscience
Primary
5029
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only