Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-P2Y2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide made to a C-terminal portion of the human P2Y2 protein (between animo acids 300-377) [UniProt P41231].
Antigen P2Y2
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11039032
View Documents
Novus Biologicals
NB11039032
0.1 mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
NB11039032T
View Documents
Novus Biologicals
NB11039032SS
0.025 mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

P2Y2 Polyclonal antibody specifically detects P2Y2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Rabbit
A synthetic peptide made to a C-terminal portion of the human P2Y2 protein (between animo acids 300-377) [UniProt P41231].
Polyclonal
Primary
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
5029
1 mg/mL
IgG
P2Y2
Affinity Purified
RUO
P41231
PBS with 0.05% Sodium Azide
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit