Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2Y6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA534093
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Monkey, Marmoset, Rat, Hamster, Bovine, Rabbit, Pig (100%).
P2Y6 Polyclonal antibody specifically detects P2Y6 in Bovine, Hamster, Human, Non-human Primate, Porcine, Rabbit samples. It is validated for Immunohistochemistry (Paraffin).Specifications
P2Y6 | |
Polyclonal | |
Unconjugated | |
P2ry6 | |
2010204J23Rik; G-coupled nucleotide receptor; MGC15335; P2 purinoceptor; P2ry6; P2Y ATP receptor 6; P2Y purinoceptor 6; p2y3 and chick; p2y3 receptor; P2Y6; P2Y6 receptor; PP2891; Purinoceptor p2y6; pyrimidinergic receptor P2Y, G-protein coupled, 6; pyrimidinergic receptor P2Y6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100358580, 100512849, 100752781, 5031, 539703 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q15077 | |
P2ry6 | |
Synthetic 17 amino acid peptide from 2nd cytoplasmic domain of human P2RY6 / P2Y6. | |
50 μg | |
Primary | |
Bovine, Hamster, Human, Non-human Primate, Porcine, Rabbit | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction