Learn More
p36/MAT1 Mouse, Unlabeled, Clone: 6, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 610532
Catalog No. BDB610532
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Regulation of the cell cycle by cyclin-dependent kinases (cdks) is controlled by the phosphorylation of a conserved threonine residue in their catalytic subunit. Cdks are activated as a result of the phosphorylation of another cyclin-dependent kinase, cdk-activating kinase (CAK). This kinase has two subunits: the catalytic Cdk7 and the regulatory cyclin H. A 36 kDa protein, p36 (also known as Menage A Trois-1 (MAT1)), coimmunoprecipitates with CAK. p36 is a member of the RING finger family of proteins characterized by a C3HC4 zinc-binding domain. p36 functions as a CAK assembly factor and is involved in the regulation of CAK activation.
Specifications
Specifications
p36/MAT1
6
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Monoclonal
Immunofluorescence, Immunoprecipitation, Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse
IgG1
RUO
Primary
Canine, Chicken, Frog, Human, Murine, Rat