Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-p44 MAP Kinase/ERK1/MAPK3 Clone: 3H4L13, Invitrogen™

p44 MAP Kinase/ERK1/MAPK3 Monoclonal antibody 701183 from Thermo Scientific™ Pierce™ specifically detects p44 MAP Kinase/ERK1/MAPK3 in Human samples, and it is validated for WB, IF, ICC and FACS

Manufacturer:  Invitrogen 701183

Encompass

Catalog No. 70-118-3


Add to cart

Description

Description

ERK-1 is a classical MAP kinase involved in intracellular signaling in response to extracellular stimulus. ERK-1 is activated by growth factors, hormones, and phorbol esters. ERKs are widely expressed in different cells, and are involved in control of cell proliferation, cell differentiation, and cytoskeletal regulation. ERK-1 is a serine/threonine kinase which phosphorylates cytoplasmic signaling molecules, cell surface receptors, transcription factors, and microtubule associated proteins upon activation by molecules upstream in the signal transduction pathway.

701183 detects p44 MAP Kinase/ERK1/MAPK3 in Human samples. 701183 has been successfully used in WB, IF, ICC and FACS procedures. The immunogen for 701183 is proprietary.

Applications:

FACS, Immunocytochemistry, Immunofluorescence, Western Blotting

Specifications

Specifications

p44 MAP Kinase/ERK1/MAPK3
Unconjugated
P27361
Rabbit
IgG
Store at 2 to 8°C for up to 1 month. For long term storage, store at -20°C.
Monoclonal
Human
3H4L13
100μg of Protein A antibody in PBS with 0.09% sodium azide;
Purified
proprietary
Protein A
Primary
5595
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit