Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p45/SUG1 Mouse, Unlabeled, Clone: 35, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611066

Catalog No. BDB611066


Add to cart

Description

Description

The 26S proteasome, a eukaryotic ATP-dependent protease complex, is responsible for selective degradation of malformed proteins and certain short-lived proteins. The 26S proteasome complex consists of the 20S proteasome (multifunctional protease), which is the catalytic core, and an ATPase-containing P700 regulatory complex. This complex contains five highly related putative ATPases (TBP1, TBP7, S4, MSS1, p45) that belong to the ATPase family designated AAA (ATPases Associated with a variety of cellular Activities). They share a highly conserved 200 amino acid ATPase domain (AAA module) and participate in a range of cellular functions. p45 (Trip1, hSUG1) is the homolog of the S. cerevisiae and mouse SUG1 proteins. In a ligand-enhanced manner, mouse SUG1 interacts with a range of nuclear receptors via their AF-2 domains and yeast SUG1 functions as a transcription factor that mediates the transcriptional response of the GAL4 protein. p45 is thought to be their functional homolog. Thus, in addition to its participation in the P700 regulatory complex, p45 may function as a mammalian transcriptional modulator.

Immunofluorescence, Western Blotting

Specifications

Specifications

p45/SUG1
35
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
SUG1
Affinity Purified
Human p45/SUG1 aa. 47-168
50μg
Cell Biology
Primary
Canine, Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit