Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p53 Mouse anti-Human, Clone: BP53-12, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05224
Description
Anti-p53 Antibody, clone BP53-12 is a high quality Mouse Monoclonal Antibody for the detection of p53 and has been validated in immunohistochemistry, immunoprecipitation and western blotting.Specifications
p53 | |
Monoclonal | |
Unconjugated | |
NM_000546 | |
Mouse | |
Protein A purfied | |
Primary | |
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles. | |
IgG2a |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
BP53-12 | |
Protein G Purified immunoglobulin in Protein G Purified immunoglobulin in of 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30% | |
TP53, P53, TRP53, p53, LFS1 | |
Recombinant human wild type p53 | |
200 μL | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only