Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p53DINP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA130836

Catalog No. PA130836


Add to cart

Description

Description

PA1-30836 detects p53 DINP1 from human, rat, mouse samples. PA1-30836 has been successfully used in Western blot applications. The PA1-30836 immunogen is: Synthetic peptide: amino acids 1-14 of human p53DINP1. This sequence is identical between alpha and beta, and differs by one amino acid from those of mouse. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

p53DINP1
Polyclonal
Unconjugated
TP53INP1
Liquid
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
297822, 60599, 94241
Western Blot
1 mg/mL
PBS with 0.02% sodium azide; pH 7.2
Q96A56, Q9QXE4, Q80YE2
TP53INP1, TRP53INP1
synthetic peptide amino acids 1-14 of human p53DINP1. This sequence is identical between alpha and beta, and differs by one amino acid from those of mouse.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit