Learn More
IFIT1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA3848
Catalog No. PA3848
Add to cart
Description
Description
PA3-848 detects ISG56 (p56) from human and mouse samples. PA3-848 has been successfully used in Western blot and ICC/IF applications to detect a 39-58 kDa band.
Specifications
Specifications
IFIT1
Polyclonal
whole serum with 0.05% sodium azide
P09914, Q64282
P56, IFIT1
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
IFIT1
Liquid
IFIT1
Bacterially expressed full-length human p56 protein
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
15957, 3434