Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p57 Kip2 Rabbit anti-Human, Polyclonal, Invitrogen™

Catalog No. PIPA129191
Change view
Click to view available options
Quantity:
500 μL
Catalog No. Quantity
PIPA129191 500 μL
1 options

Catalog No. PIPA129191

Supplier: Thermo Scientific PA129191

Rabbit Polyclonal Antibody

PA1-29194 detects PI 3 Kinase p85 from non-human primate, rat, human samples. PA1-29194 has been successfully used in Western blot applications. The PA1-29194 immunogen is: Synthetic peptide from the C terminal SH2 domain of p85 alpha subunit of phospha

TRUSTED_SUSTAINABILITY

Specifications

Antigen p57 Kip2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Gene CDKN1C
Gene Accession No. P49918
Gene Symbols CDKN1C
Host Species Rabbit
Immunogen Synthetic peptide from human p57Kip2 protein.
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1028
Target Species Human
Content And Storage 4° C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.